missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfate Buffer, Reagents

Catalog No. 01333913
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01333913 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01333913 Supplier Labchem Inc Supplier No. CS0231001A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sodium Sulfate Buffer, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Buffer
CAS 7732-18-5
Color Colorless
Physical Form Liquid
Quantity 1 L
Packaging Poly Bottle
Sterility Non-sterile
Solubility Information Completely miscible