missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Sulfate Anhydrous, min.99%, 12-60 Mesh, Granular, Ultra Resi-Analyzed, ACS Reagent, J.T. Baker™
$2225.44 - $2225.44
Chemical Identifiers
| CAS | 7757-82-6 |
|---|---|
| Molecular Formula | Na2O4S |
| Molecular Weight (g/mol) | 142.04 |
| MDL Number | MFCD00003504 |
| InChI Key | PMZURENOXWZQFD-UHFFFAOYSA-L |
| Synonym | sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1 |
| PubChem CID | 24436 |
| ChEBI | CHEBI:32149 |
| SMILES | [Na+].[Na+].[O-]S([O-])(=O)=O |
Chemical Identifiers
| 7757-82-6 | |
| 142.04 | |
| PMZURENOXWZQFD-UHFFFAOYSA-L | |
| 24436 | |
| [Na+].[Na+].[O-]S([O-])(=O)=O |
| Na2O4S | |
| MFCD00003504 | |
| sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1 | |
| CHEBI:32149 |
Specifications
| 7757-82-6 | |
| Granules | |
| Na2O4S | |
| sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1 | |
| [Na+].[Na+].[O-]S([O-])(=O)=O | |
| 142.04 | |
| CHEBI:32149 | |
| Meets ACS reagent chemical requirements | |
| Wide Mouth Amber Glass Bottle | |
| Sodium Sulfate, Anhydrous |
| 5.2 to 9.2 | |
| 500 g | |
| MFCD00003504 | |
| PMZURENOXWZQFD-UHFFFAOYSA-L | |
| disodium;sulfate | |
| 24436 | |
| 142.04 | |
| max. 0.5% | |
| 1L = 2.68kg |