missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfate Anhydrous (Low Nitrogen), Fisher Chemical™

Catalog No. p-34320
Change view
Click to view available options
Quantity:
3 kg
10 kg
Packaging:
Glass Bottle
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S4203 3 kg Glass Bottle
S42010 10 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S4203 Supplier Thermo Fisher Scientific Supplier No. S420 3
Only null left

Chemical Identifiers

CAS 7757-82-6
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
Synonym sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
PubChem CID 24436
ChEBI CHEBI:32149
IUPAC Name disodium sulfate
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O

Specifications

Melting Point 884°C
Color White
pH 5 to 8
Physical Form Solid
Quantity 3 kg
Formula Weight 142.04
Identification Pass Test
Packaging Glass Bottle
Decomposition Information Decomposition Information: >890°C
Chemical Name or Material Sodium Sulfate Anhydrous
Safety handling Check Alert storage code Red
Hazard Category SIGNAL WORD: CAUTION!
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause skin, eye, and respiratory tract irritation. The toxicological properties have not been fully investigated. Hygroscopic.
NFPA
Health: 1
Flammability: 1
Instability: 0