missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Sulfate, Anhydrous, Granular Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Specifications
Specifications
| Physical Form | Granular Powder |
| Quantity | 2.5 kg |
| Formula Weight | 142.04 |
| Percent Purity | 99.8% |
| Grade | BAKER ANALYZED™ ACS Reagent |
| Packaging | Wide Mouth Amber Glass Bottle |
| Specific Gravity | 1L = 2.68kg |
| Chemical Name or Material | Sodium Sulfate, Anhydrous |