missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Sulfate Anhydrous (Granular/Certified ACS), Fisher Chemical™
Supplier: Thermo Fisher Scientific S42150
Specifications
| Sodium Sulfate Anhydrous | |
| 7757-82-6 | |
| 5 to 8 | |
| 50 kg | |
| ≥99.0% | |
| MFCD00003504 | |
| PMZURENOXWZQFD-UHFFFAOYSA-L | |
| disodium sulfate | |
| 24436 | |
| 142.04 | |
| Certified ACS | |
| 0.5% max. | |
| Decomposition Information: >890°C |
| 884°C | |
| White | |
| Solid | |
| ≥99.0 % | |
| Na2O4S | |
| sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1 | |
| [Na+].[Na+].[O-]S([O-])(=O)=O | |
| 142.04 | |
| CHEBI:32149 | |
| ≥99.0% | |
| Pass Test | |
| Fiber Drum |
Chemical Identifiers
| 7757-82-6 | |
| 142.04 | |
| PMZURENOXWZQFD-UHFFFAOYSA-L | |
| 24436 | |
| disodium sulfate |
| Na2O4S | |
| MFCD00003504 | |
| sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1 | |
| CHEBI:32149 | |
| [Na+].[Na+].[O-]S([O-])(=O)=O |
Safety and Handling
CAUTION!
Emergency Overview
May cause skin, eye, and respiratory tract irritation. The toxicological properties have not been fully investigated. Hygroscopic.
NFPA
Health: 1
Flammability: 1
Instability: 0
SIGNAL WORD: CAUTION!