missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Sulfate Anhydrous, Certified ACS, Suitable for Pesticide Residue Analysis (Granular, 10-60 mesh), Fisher Chemical™
Specifications
Specifications
| Melting Point | 884°C |
| Color | White |
| pH | 5 to 8 |
| Physical Form | Solid |
| Quantity | 2.5 kg |
| Identification | Pass Test |
| Loss on Ignition | 0.5% max. |
| Packaging | Glass Bottle |
| Decomposition Information | Decomposition Information: >890°C |
| Assay Percent Range | ≥99.0 % |
| Show More |
Safety and Handling
| Hazard Category | SIGNAL WORD: CAUTION! |
| Hazard Statement | CAUTION! |
| Precautionary Statement | Emergency Overview May cause skin, eye, and respiratory tract irritation. The toxicological properties have not been fully investigated. Hygroscopic. NFPA Health: 1 Flammability: 1 Instability: 0 |