missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Sulfate Anhydrous, ACS Reagent, Low Organic, Reagents
Specifications
Specifications
| Boiling Point | >890°C |
| Melting Point | 884°C to 888°C |
| Color | Colorless/White |
| Physical Form | Solid |
| Quantity | 12 kg |
| Grade | ACS |
| Packaging | Poly Pail |
| Specific Gravity | 2.7 |
| Chemical Name or Material | Sodium Sulfate Anhydrous, ACS Reagent, Low Organic |