missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Sulfate Anhydrous, ACS Reagent, Low Organic, Reagents
Sodium Sulfate Anhydrous, ACS Reagent, Low Organic, ACS, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C132555025KG
Specifications
| >890°C | |
| 884°C to 888°C | |
| 7757-82-6 | |
| 2.5 kg | |
| MFCD00003504 | |
| PMZURENOXWZQFD-UHFFFAOYSA-L | |
| disodium sulfate | |
| ACS | |
| 2.7 |
| Sodium Sulfate Anhydrous, ACS Reagent, Low Organic | |
| Colorless/White | |
| Solid | |
| Na2O4S | |
| Sulfuric Acid disodium salt | |
| [Na+].[Na+].[O-]S([O-])(=O)=O | |
| 142.04 | |
| Glass Bottle |
Chemical Identifiers
| 7757-82-6 | |
| 142.04 | |
| PMZURENOXWZQFD-UHFFFAOYSA-L | |
| disodium sulfate |
| Na2O4S | |
| MFCD00003504 | |
| Sulfuric Acid disodium salt | |
| [Na+].[Na+].[O-]S([O-])(=O)=O |