missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Sulfate Anhydrous, ACS Reagent, Low Organic, Reagents
Sodium Sulfate Anhydrous, ACS Reagent, Low Organic, ACS, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C132555050F4
Specifications
| >890°C | |
| 884°C to 888°C | |
| 7757-82-6 | |
| 50 kg | |
| MFCD00003504 | |
| PMZURENOXWZQFD-UHFFFAOYSA-L | |
| disodium sulfate | |
| ACS | |
| 2.7 |
| Sodium Sulfate Anhydrous, ACS Reagent, Low Organic | |
| Colorless/White | |
| Solid | |
| Na2O4S | |
| Sulfuric Acid disodium salt | |
| [Na+].[Na+].[O-]S([O-])(=O)=O | |
| 142.04 | |
| Fiber Drum |
Chemical Identifiers
| 7757-82-6 | |
| 142.04 | |
| PMZURENOXWZQFD-UHFFFAOYSA-L | |
| disodium sulfate |
| Na2O4S | |
| MFCD00003504 | |
| Sulfuric Acid disodium salt | |
| [Na+].[Na+].[O-]S([O-])(=O)=O |