missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfate, Anhydrous, ACS Grade, ≥ 99.0%, LabChem™

Catalog No. p-5388255
Change view
Click to view available options
Quantity:
500 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC248801 500 g
LC248802 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC248801 Supplier LabChem Supplier No. LC248801

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7757-82-6
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
Synonym sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
PubChem CID 24436
ChEBI CHEBI:32149
IUPAC Name disodium sulfate
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O

Specifications

Melting Point 884°C
Color White
CAS Max % 100
Physical Form Crystals
Quantity 500 g
Linear Formula Na2SO4
Solubility Information Soluble in water
Formula Weight 142.04
Percent Purity ≥99.0%
Grade ACS
Identification Passes Test
Packaging Poly Bottle
Decomposition Information Sulfur compounds
Chemical Name or Material Sodium Sulfate, Anhydrous
Show More Show Less
Hazard Statement GHS H Statement
Product is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
EINECSNumber 231-820-9
Recommended Storage Room Temperature