missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulfate, anhydrous, 99%

Catalog No. AAA198900B
Change view
Click to view available options
Quantity:
1000 g
5000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA198900B 1000 g
AAA198900I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA198900B Supplier Thermo Scientific Chemicals Supplier No. A198900B

CAS: 7757-82-6 | Na2O4S | 142.04 g/mol

Sodium sulfate anhydrous is used primarily for drying non-aqueous solvents by removing traces of water. It is used as a fining agent for molten glass, levelling agent in textile processing, and filler in detergents. It is also used in Kraft process for paper pulping, defrosting windows, manufacture of starch, glass, and soap.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium sulfate anhydrous is used primarily for drying non-aqueous solvents by removing traces of water. It is used as a fining agent for molten glass, levelling agent in textile processing, and filler in detergents. It is also used in Kraft process for paper pulping, defrosting windows, manufacture of starch, glass, and soap.

Solubility
Soluble in water, glycerol and hydrogen iodide. Insoluble in ethanol and many organic solvents.

Notes
Anhydrous sodium sulfate is hygroscopic in nature, and absorbs moisture converting to the hydrated form. The hydrated form on heating gets converted back to anhydrous form.

Chemical Identifiers

CAS 7757-82-6
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
Synonym sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
PubChem CID 24436
ChEBI CHEBI:32149
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O

Specifications

Melting Point 884°C
Quantity 1000 g
Linear Formula Na2SO4
Merck Index 14,8680
Solubility Information Soluble in water,glycerol and hydrogen iodide. Insoluble in ethanol and many organic solvents.
IUPAC Name disodium;sulfate
Formula Weight 142.04
Percent Purity 99%
Odor Odorless
Refractive Index 1.484
Sensitivity Hygroscopic
Density 2.68
Chemical Name or Material Sodium sulfate, anhydrous
Show More Show Less
EINECSNumber 231-820-9
RTECSNumber WE1650000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only