missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfate, Anhydrous , 99.0% min., ACS, MilliporeSigma™

Catalog No. MSX07605
Change view
Click to view available options
Quantity:
12 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MSX07605 12 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MSX07605 Supplier Technidata Supplier No. SX07605

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ACS reagent grade anhydrous sodium sulfate suitable for analytical and research applications.

Chemical Identifiers

CAS 7757-82-6
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
Synonym sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
PubChem CID 24436
ChEBI CHEBI:32149
IUPAC Name disodium sulfate
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O

Specifications

Color White
Physical Form Granules
Quantity 12 kg
Assay Percent Range 99.0% min.
Formula Weight 142.04
Percent Purity ≥99.0%
Grade ACS
Chemical Name or Material Sodium Sulfate, Anhydrous, For ACS analysis