missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Sulfate, ACS Reagent, Anhydrous Powder, Honeywell Fluka™
ACS Reagent, anhydrous, powder,≥99.0%
$182.87 - $659.82
Chemical Identifiers
| CAS | 7757-82-6 |
|---|---|
| Molecular Formula | Na2O4S |
| Molecular Weight (g/mol) | 142.04 |
| MDL Number | MFCD00003504 |
| InChI Key | PMZURENOXWZQFD-UHFFFAOYSA-L |
| Synonym | sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1 |
| PubChem CID | 24436 |
| ChEBI | CHEBI:32149 |
| IUPAC Name | disodium;sulfate |
| SMILES | [Na+].[Na+].[O-]S([O-])(=O)=O |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 7757-82-6 | |
| 142.04 | |
| PMZURENOXWZQFD-UHFFFAOYSA-L | |
| 24436 | |
| disodium;sulfate |
| Na2O4S | |
| MFCD00003504 | |
| sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1 | |
| CHEBI:32149 | |
| [Na+].[Na+].[O-]S([O-])(=O)=O |
Specifications
| 7757-82-6 | |
| 500 g | |
| Na2SO4 | |
| NONH for all modes of transport | |
| PMZURENOXWZQFD-UHFFFAOYSA-L | |
| disodium;sulfate | |
| 24436 | |
| 142.04g/mol | |
| ACS Reagent | |
| Sodium Sulfate |
| Anhydrous Powder | |
| Na2O4S | |
| MFCD00003504 | |
| sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1 | |
| [Na+].[Na+].[O-]S([O-])(=O)=O | |
| 142.04 | |
| CHEBI:32149 | |
| ≥99% | |
| 2.68g/mL at 25°C |
Safety and Handling
EINECSNumber : 231-820-9
RTECSNumber : WE1650000