missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Sulfate, ACS Reagent, Anhydrous Granular, Honeywell Fluka™
ACS Reagent, anhydrous, granular,≥99.0%
Supplier: Honeywell-Fluka 2393131KG
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
| Sodium Sulfate | |
| Granules | |
| Na2O4S | |
| MFCD00003504 | |
| sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1 | |
| [Na+].[Na+].[O-]S([O-])(=O)=O | |
| 142.04 | |
| CHEBI:32149 | |
| ≥99% | |
| 2.68g/mL at 25°C |
| 7757-82-6 | |
| 1 kg | |
| Na2SO4 | |
| NONH for all modes of transport | |
| PMZURENOXWZQFD-UHFFFAOYSA-L | |
| disodium;sulfate | |
| 24436 | |
| 142.04g/mol | |
| ACS Reagent |
Chemical Identifiers
| 7757-82-6 | |
| 142.04 | |
| PMZURENOXWZQFD-UHFFFAOYSA-L | |
| 24436 | |
| [Na+].[Na+].[O-]S([O-])(=O)=O |
| Na2O4S | |
| MFCD00003504 | |
| sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1 | |
| CHEBI:32149 |
Safety and Handling
EINECSNumber : 231-820-9
RTECSNumber : WE1650000