missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulfate, 99%, extra pure, anhydrous

Catalog No. p-3523586
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 kg
2.5 kg
25 g
5 kg
Packaging:
Glass Bottle
Plastic bottle
Catalog No. Quantity Packaging
AC196640250 25 g Glass Bottle
AC196640010 1 kg Plastic bottle
AC196640025 2.5 kg Plastic bottle
AC196640050 5 kg Plastic bottle
4 options

Catalog No. AC196640250

Supplier: Thermo Scientific Chemicals 196640250

Only null left

CAS: 7757-82-6 | Na2O4S | 142.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7757-82-6
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
Synonym sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
PubChem CID 24436
ChEBI CHEBI:32149
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O

Specifications

Boiling Point >1700.0°C
Melting Point 884.0°C
Color White
Physical Form Crystalline Powder
Quantity 25 g
Assay Percent Range 98.5% min
Linear Formula Na2SO4
Fieser 01,1103
Merck Index 15, 8809
Solubility Information Solubility in water: 185g/L. Other solubilities: soluble in glycerol and hydrogen iodide, insoluble in alcohol
IUPAC Name disodium;sulfate
Formula Weight 142.04
Percent Purity 99%
Grade Extra Pure
Packaging Glass Bottle
Density 2.68
Chemical Name or Material Sodium sulfate
Show More Show Less
EINECSNumber 231-820-9

RUO – Research Use Only