missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sodium sulfate, 99%, for biochemistry, anhydrous

Catalog No. p-4813665
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
500 g
Catalog No. Quantity
AC447925000 500 g
1 options

Catalog No. AC447925000

Supplier: Thermo Scientific Chemicals 447925000

Only null left

CAS: 7757-82-6 | Na2O4S | 142.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7757-82-6
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
Synonym sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
PubChem CID 24436
ChEBI CHEBI:32149
IUPAC Name disodium;sulfate
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O

Specifications

Boiling Point >1700.0°C
Melting Point 884.0°C
Quantity 500 g
Assay Percent Range 99%
Linear Formula Na2SO4
Merck Index 15,8809
Solubility Information (10% in water) Clear colorless
Formula Weight 142.04
Percent Purity 99%
Grade Biochemical
Packaging Plastic bottle
Chemical Name or Material Sodium sulfate
Show More Show Less
EINECSNumber 231-820-9

RUO – Research Use Only