missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulfate, 99+%, for analysis, crystalline, anhydrous

CAS: 7757-82-6 | Na2O4S | 142.04 g/mol

Supplier:  Thermo Scientific Chemicals 219260025

 View more versions of this product

Catalog No. AC219260025


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

>1700.0°C
884.0°C
7757-82-6
5 to 8 (5% soln. at 20°C)
2.5 kg
Na2O4S
MFCD00003504
15, 8809
Solubility in water: 185g/L. Other solubilities: soluble in glycerol and hydrogen iodide, insoluble in alcohol
[Na+].[Na+].[O-]S([O-])(=O)=O
142.04
CHEBI:32149
≥99%
0.5% max.
2.68g/mL
Sodium sulfate
Anhydrous
White
Crystalline Powder and/or Crystals
99+%
Na2SO4
01,1103
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
PMZURENOXWZQFD-UHFFFAOYSA-L
disodium;sulfate
24436
142.04
Analytical
Plastic bottle

Chemical Identifiers

7757-82-6
142.04
PMZURENOXWZQFD-UHFFFAOYSA-L
24436
disodium;sulfate
Na2O4S
MFCD00003504
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
CHEBI:32149
[Na+].[Na+].[O-]S([O-])(=O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-820-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only