missing translation for 'onlineSavingsMsg'
Learn More

Sodium Succinate, Hexahydrate, Reagent, 97-102%, Spectrum™ Chemical

Catalog No. 18606812
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606812 100 g Amber Glass Bottle
18606813 500 g Amber Glass Bottle
18606814 2.5 kg Poly Pail
18606815 12 kg Fiber Drum
18606816 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18606812 Supplier Spectrum Chemical Mfg Cor Supplier No. S1449100GM
Get Up to $500 in Chemicals with Purchase
View all promotions

S1449, 6106-21-4, C4H4Na2O4·6H2O

Spectrum™ Chemical Sodium Succinate, Hexahydrate, Reagent , also known as succinic acid, is a diprotic and dicarboxylic acid. It is involved in an energy-yielding process known as the citric acid cycle. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 6106-21-4
Molecular Formula C4H16Na2O10
Molecular Weight (g/mol) 270.14
InChI Key ZBTUYCUNQBRXOR-UHFFFAOYSA-L
IUPAC Name disodium butanedioate hexahydrate
SMILES O.O.O.O.O.O.[Na+].[Na+].[O-]C(=O)CCC([O-])=O

Specifications

CAS Max % 1
Quantity 100 g
Percent Purity 97 to 102%
Grade Reagent
Packaging Amber Glass Bottle