missing translation for 'onlineSavingsMsg'
Learn More

Sodium Succinate Granular, Macron Fine Chemicals™

Catalog No. MK7980500
Change view
Click to view available options
Quantity:
500 g
Packaging:
POLYSTORMOR
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MK7980500 500 g POLYSTORMOR
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK7980500 Supplier Avantor Supplier No. 798003

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6106-21-4
Molecular Formula C4H16Na2O10
Molecular Weight (g/mol) 270.142
InChI Key ZBTUYCUNQBRXOR-UHFFFAOYSA-L
Synonym disodium succinate hexahydrate, sodium succinate hexahydrate, unii-u16qod6c4e, butanedioic acid, disodium salt, hexahydrate, disodium butanedioate hexahydrate, u16qod6c4e, disodium hexahydrate succinate, sodium succinate dibasic hexahydrate, butanedioic acid disodium salt, succinic acid disodium salt hexahydrate
PubChem CID 3083938
ChEBI CHEBI:63686
IUPAC Name disodium;butanedioate;hexahydrate
SMILES C(CC(=O)[O-])C(=O)[O-].O.O.O.O.O.O.[Na+].[Na+]

Specifications

Packaging POLYSTORMOR
Quantity 500 g
Formula Weight 270.16
Physical Form Granules
Chemical Name or Material Sodium Succinate