missing translation for 'onlineSavingsMsg'
Learn More

Sodium Succinate, Anhydrous, Reagent, 99-102%, Spectrum™ Chemical

Catalog No. 18606808
Change view
Click to view available options
Quantity:
100 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606808 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18606808 Supplier Spectrum Chemical Mfg Cor Supplier No. S1361100GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

S1361, 150-90-3, C4H4Na2O4

Spectrum™ Chemical Sodium Succinate, Anhydrous, Reagent is the sodium salt of a dicarboxylic acid which is used primarily as an acidity regulator and an industrial biotechnology raw material. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 150-90-3
Molecular Formula C4H4Na2O4
Molecular Weight (g/mol) 162.05
InChI Key ZDQYSKICYIVCPN-UHFFFAOYSA-L
IUPAC Name disodium butanedioate
SMILES [Na+].[Na+].[O-]C(=O)CCC([O-])=O

Specifications

CAS Max % 1
Loss on Drying 0.01
Packaging Amber Glass Bottle
Quantity 100 g
Percent Purity 99 to 102%
Grade Reagent