missing translation for 'onlineSavingsMsg'
Learn More

Sodium Succinate, Anhydrous, Reagent, 99-102%, Spectrum™ Chemical

Catalog No. 18606811
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Amber Glass Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606811 12 kg Poly Pail
18606809 500 g Amber Glass Bottle
18606810 2.5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18606811 Supplier Spectrum Chemical Mfg Cor Supplier No. S136112KG
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

S1361, 150-90-3, C4H4Na2O4

Spectrum™ Chemical Sodium Succinate, Anhydrous, Reagent is the sodium salt of a dicarboxylic acid which is used primarily as an acidity regulator and an industrial biotechnology raw material. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 150-90-3
Molecular Formula C4H4Na2O4
Molecular Weight (g/mol) 162.05
InChI Key ZDQYSKICYIVCPN-UHFFFAOYSA-L
IUPAC Name disodium butanedioate
SMILES [Na+].[Na+].[O-]C(=O)CCC([O-])=O

Specifications

CAS Max % 1
Loss on Drying 0.01
Packaging Poly Pail
Quantity 12 kg
Percent Purity 99 to 102%
Grade Reagent