missing translation for 'onlineSavingsMsg'
Learn More

Sodium sticks, coated in film of protective hydrocarbon oil, 99%

Catalog No. AAL1328524
Change view
Click to view available options
Quantity:
125 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1328524 125 g
AAL1328536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1328524 Supplier Thermo Scientific Chemicals Supplier No. L1328524

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7440-23-5 | Na | 22.99 g/mol

Sodium sticks, in mineral oil is used for reviews of Na metal - ammonia (Birch) reductions, for preparation of sodamide in liquid ammonia and for use in an acyloin synthesis promoted by sonication.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium sticks, in mineral oil is used for reviews of Na metal - ammonia (Birch) reductions, for preparation of sodamide in liquid ammonia and for use in an acyloin synthesis promoted by sonication.

Solubility
Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Handle and store under inert gas. It is sensitive to air.

Chemical Identifiers

CAS 7440-23-5
Molecular Formula Na
Molecular Weight (g/mol) 22.99
MDL Number MFCD00085307
InChI Key KEAYESYHFKHZAL-UHFFFAOYSA-N
Synonym natrium, sodium-23, sodio, metal, sodio spanish, liquid alloy, unii-9nez333n27, hsdb 687, unii-23j3bhr95o, ingot
PubChem CID 5360545
ChEBI CHEBI:26708
IUPAC Name sodium
SMILES [Na]

Specifications

Melting Point 98°C
Boiling Point 881°C
Quantity 125 g
Flash Point >110°C
Sensitivity Air and moisture sensitive
UN Number UN1428
Merck Index 14,8570
Solubility Information Reacts with water.
Percent Purity 99%
Chemical Name or Material Sodium sticks, in mineral oil
Hazard Category H260-H304-H314-H335
Hazard Statement GHS H Statement

H260-H314

In contact with water releases flammable gases which may ignite spontaneously.

Causes severe skin burns and eye damage.
Precautionary Statement P223-P231+P232-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: SODIUM
EINECSNumber 231-132-9
RTECSNumber VY0686000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only