missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Stearyl Fumarate, NF, 99-101.5%, Spectrum™ Chemical
S1448, 4070-80-8, C22H39NaO4
$5216.67 - $89746.00
Chemical Identifiers
| CAS | 4070-80-8 |
|---|---|
| Molecular Formula | C22H39NaO4 |
| Molecular Weight (g/mol) | 390.54 |
| InChI Key | STFSJTPVIIDAQX-LTRPLHCISA-M |
| IUPAC Name | sodium (2E)-4-(octadecyloxy)-4-oxobut-2-enoate |
| SMILES | [Na+].CCCCCCCCCCCCCCCCCCOC(=O)\C=C\C([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18606806
|
Spectrum Chemical Mfg Cor
S1448500GM |
500 g | Poly Pail |
Each for $5,216.67
|
|
||||
|
18606807
|
Spectrum Chemical Mfg Cor
S144825KGBL |
25 kg | Poly Pail |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Sodium Stearyl Fumarate, NF is a water-soluble tablet lubicant. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 4070-80-8 | |
| 390.54 | |
| sodium (2E)-4-(octadecyloxy)-4-oxobut-2-enoate |
| C22H39NaO4 | |
| STFSJTPVIIDAQX-LTRPLHCISA-M | |
| [Na+].CCCCCCCCCCCCCCCCCCOC(=O)\C=C\C([O-])=O |
Specifications
| 4070-80-8 | |
| > 200°C | |
| C22H39NaO4 | |
| 142.2-146.0 | |
| [Na+].CCCCCCCCCCCCCCCCCCOC(=O)\C=C\C([O-])=O | |
| 390.54 | |
| NF | |
| Sodium Stearyl Fumarate |
| 1 | |
| Poly Pail | |
| 500 g | |
| STFSJTPVIIDAQX-LTRPLHCISA-M | |
| sodium (2E)-4-(octadecyloxy)-4-oxobut-2-enoate | |
| 99 to 101.5% | |
| Crystalline Powder or Lumps |