missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Stearyl Fumarate, NF, 99-101.5%, Spectrum™ Chemical
S1448, 4070-80-8, C22H39NaO4
$625.24 - $1806.82
Chemical Identifiers
| CAS | 4070-80-8 |
|---|---|
| Molecular Formula | C22H39NaO4 |
| Molecular Weight (g/mol) | 390.54 |
| InChI Key | STFSJTPVIIDAQX-LTRPLHCISA-M |
| IUPAC Name | sodium (2E)-4-(octadecyloxy)-4-oxobut-2-enoate |
| SMILES | [Na+].CCCCCCCCCCCCCCCCCCOC(=O)\C=C\C([O-])=O |
Description
Spectrum™ Chemical Sodium Stearyl Fumarate, NF is a water-soluble tablet lubicant. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 4070-80-8 | |
| 390.54 | |
| sodium (2E)-4-(octadecyloxy)-4-oxobut-2-enoate |
| C22H39NaO4 | |
| STFSJTPVIIDAQX-LTRPLHCISA-M | |
| [Na+].CCCCCCCCCCCCCCCCCCOC(=O)\C=C\C([O-])=O |
Specifications
| 4070-80-8 | |
| White | |
| C22H39NaO4 | |
| 142.2-146 | |
| [Na+].CCCCCCCCCCCCCCCCCCOC(=O)\C=C\C([O-])=O | |
| 390.54 | |
| NF | |
| Sodium Stearyl Fumarate |
| 1 | |
| Poly Bottle | |
| 25 g | |
| STFSJTPVIIDAQX-LTRPLHCISA-M | |
| sodium (2E)-4-(octadecyloxy)-4-oxobut-2-enoate | |
| 99 to 101.5% | |
| Crystalline Powder or Lumps |