missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Sorbate 98.0+%, TCI America™
$119.36 - $1326.44
Chemical Identifiers
| CAS | 7757-81-5 |
|---|---|
| Molecular Formula | C6H7NaO2 |
| Molecular Weight (g/mol) | 134.11 |
| MDL Number | MFCD00058995 |
| InChI Key | LROWVYNUWKVTCU-STWYSWDKSA-M |
| Synonym | Sorbic Acid Sodium Salt |
| PubChem CID | 23665582 |
| IUPAC Name | sodium;(2E,4E)-hexa-2,4-dienoate |
| SMILES | CC=CC=CC(=O)[O-].[Na+] |
Chemical Identifiers
| 7757-81-5 | |
| 134.11 | |
| LROWVYNUWKVTCU-STWYSWDKSA-M | |
| 23665582 | |
| CC=CC=CC(=O)[O-].[Na+] |
| C6H7NaO2 | |
| MFCD00058995 | |
| Sorbic Acid Sodium Salt | |
| sodium;(2E,4E)-hexa-2,4-dienoate |
Specifications
| White-Yellow | |
| Crystalline Powder | |
| C6H7NaO2 | |
| Sorbic Acid Sodium Salt | |
| CC=CC=CC(=O)[O-].[Na+] | |
| 134.11 | |
| 134.11 | |
| Sodium Sorbate |
| 7757-81-5 | |
| 25 g | |
| MFCD00058995 | |
| LROWVYNUWKVTCU-STWYSWDKSA-M | |
| sodium;(2E,4E)-hexa-2,4-dienoate | |
| 23665582 | |
| ≥98.0% (T) |
Safety and Handling
EINECSNumber : (2)-0611
RTECSNumber : WG2220000
TSCA : No