missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Silicate, 40 DEG Be Solution, Spectrum™ Chemical
Specifications
Specifications
| CAS | 6834-92-0, 7732-18-5, 1344-09-8 |
| Quantity | 500 mL |
| Grade | Reagent |
| Packaging | Amber Glass Bottle |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.