missing translation for 'onlineSavingsMsg'
Learn More

Sodium Silicate, 40 DEG Be Solution, Spectrum™ Chemical

Catalog No. 18613854
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Bottle
Poly Drum
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613854 500 mL Amber Glass Bottle
18613851 500 mL Poly Bottle
18613856 4 L Amber Glass Bottle
18613855 4 L Poly Bottle
18613852 20 L Poly Drum
18613853 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18613854 Supplier Spectrum Chemical Mfg Cor Supplier No. S1435500MLGL
Get Up to $500 in Chemicals with Purchase
View all promotions

Specifications

CAS 6834-92-0, 7732-18-5, 1344-09-8
Quantity 500 mL
Grade Reagent
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.