missing translation for 'onlineSavingsMsg'
Learn More

Sodium Selenite Pentahydrate, Solstice

Catalog No. p-7071688
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001552 25 g
6001551 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6001552 Supplier Honeywell Inc Supplier No. 0016325G

Puriss. p.a.,≥99.0% (RT)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 26970-82-1
Molecular Formula H10Na2O8Se
Molecular Weight (g/mol) 263.023
MDL Number MFCD00149173
InChI Key TUANAMBRHOLYTH-UHFFFAOYSA-L
Synonym sodium selenite pentahydrate, disodium selenite pentahydrate, unii-0wv4l961zv, ccris 1378, na2seo3.5h2o, selenious acid, disodium salt, pentahydrate, disodium pentahydrate selenite, sodium selenite hydrate 2:1:5, sodium selenite hydrate, disodium selenite hydrate
PubChem CID 134930
IUPAC Name disodium;selenite;pentahydrate
SMILES O.O.O.O.O.[O-][Se](=O)[O-].[Na+].[Na+]

Specifications

Quantity 25 g
Linear Formula Na2SeO3 · 5H2O
UN Number 2630
Formula Weight 263.01g/mol
Percent Purity ≥99% (RT)
Chemical Name or Material Sodium Selenite Pentahydrate
Hazard Statement P261-P273-P280-P301 + P310 + P330-P304 + P340 + P312-P403 + P233
Precautionary Statement H300-H317-H331-H411
EINECSNumber 233-267-9
RTECSNumber VS7420000