missing translation for 'onlineSavingsMsg'
Learn More

Sodium Selenite, 99.8+%, Powder, Spectrum™ Chemical

Catalog No. 18613849
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
5 kg
25 kg
Packaging:
Amber Glass Bottle
Lined Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613849 25 g Amber Glass Bottle
18613846 100 g Amber Glass Bottle
18613847 500 g Amber Glass Bottle
18613850 5 kg Poly Pail
18613848 25 kg Lined Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18613849 Supplier Spectrum Chemical Mfg Cor Supplier No. SO20625GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 10102-18-8
Molecular Formula Na2O3Se
Molecular Weight (g/mol) 172.95
InChI Key BVTBRVFYZUCAKH-UHFFFAOYSA-L
IUPAC Name disodium selenite
SMILES [Na+].[Na+].[O-][Se]([O-])=O

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity ≥99.8%
Grade Reagent
Packaging Amber Glass Bottle

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.