missing translation for 'onlineSavingsMsg'
Learn More

Sodium Rod, Diameter 2.5cm (Protective Liquid: Paraffin Oil), MilliporeSigma™

Catalog No. M1062600250
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1062600250 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1062600250 Supplier Technidata Supplier No. 1.06260.0250

missing translation for 'validMainframeMsg'

Specifications

CAS 7440-23-5
Appearance Silver solid
Quantity 250 g
Odor Odorless
Packaging Glass bottle
Form Solid
Molecular Formula Na
MDL Number MFCD00085307
UN Number UN 1428
Synonym natrium, sodium-23, sodio, metal, sodio spanish, liquid alloy, unii-9nez333n27, hsdb 687, unii-23j3bhr95o, ingot
InChI Key KEAYESYHFKHZAL-UHFFFAOYSA-N
SMILES [Na]
IUPAC Name sodium
Molecular Weight (g/mol) 22.99
PubChem CID 5360545
ChEBI CHEBI:26708
Formula Weight 22.99. (molar mass)
Chemical Name or Material Sodium
Show More Show Less
Hazard Category H260: In contact with water releases flammable gases which may ignite spontaneously.
H314: Causes severe skin burns and eye damage.
EUH014: Reacts violently with water.
DOTInformation DOT Class: 4.3, Packaging Group: I
EINECSNumber 231-132-9