missing translation for 'onlineSavingsMsg'
Learn More

Sodium Pyruvate 97.0+%, TCI America™

Catalog No. P0582500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0582500G 500 g
P058225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0582500G Supplier TCI America Supplier No. P0582500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 113-24-6
Molecular Formula C3H3NaO3
Molecular Weight (g/mol) 110.044
MDL Number MFCD00002586
InChI Key DAEPDZWVDSPTHF-UHFFFAOYSA-M
Synonym sodium pyruvate, pyruvic acid sodium salt, sodium 2-oxopropanoate, pyruvic acid, sodium salt, propanoic acid, 2-oxo-, sodium salt, sodium alpha-ketopropionate, pyruvate sodium, unii-pod38aif08, 2-oxopropanoic acid sodium salt, pod38aif08
PubChem CID 23662274
ChEBI CHEBI:50144
IUPAC Name sodium;2-oxopropanoate
SMILES CC(=O)C(=O)[O-].[Na+]

Specifications

Color White
Quantity 500 g
Formula Weight 110.04
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Pyruvate
EINECSNumber (9)-1079
TSCA Yes
Recommended Storage Refrigerator