missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium pyruvate, Cell Culture Grade
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White to Yellow |
| Physical Form | Powder |
| Quantity | 50 g |
| Linear Formula | CH3COCOONa |
| Beilstein | 3568341 |
| Solubility Information | Freely soluble in water |
| Formula Weight | 110.04 |
| Grade | Cell Culture |
| Chemical Name or Material | Sodium pyruvate |
Safety and Handling
| EINECSNumber | 204-024-4 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only