missing translation for 'onlineSavingsMsg'
Learn More

Sodium pyruvate, Cell Culture Grade

Catalog No. AAJ6184018
Change view
Click to view available options
Quantity:
50 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6184018 50 g
AAJ6184022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6184018 Supplier Thermo Scientific Chemicals Supplier No. J6184018

CAS: 113-24-6 | C3H3NaO3 | 110.044 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 113-24-6
Molecular Formula C3H3NaO3
Molecular Weight (g/mol) 110.044
MDL Number MFCD00002586
InChI Key DAEPDZWVDSPTHF-UHFFFAOYSA-M
Synonym sodium pyruvate, pyruvic acid sodium salt, sodium 2-oxopropanoate, pyruvic acid, sodium salt, propanoic acid, 2-oxo-, sodium salt, sodium alpha-ketopropionate, pyruvate sodium, unii-pod38aif08, 2-oxopropanoic acid sodium salt, pod38aif08
PubChem CID 23662274
ChEBI CHEBI:50144
IUPAC Name sodium;2-oxopropanoate
SMILES CC(=O)C(=O)[O-].[Na+]

Specifications

Color White to Yellow
Physical Form Powder
Quantity 50 g
Linear Formula CH3COCOONa
Beilstein 3568341
Solubility Information Freely soluble in water
Formula Weight 110.04
Grade Cell Culture
Chemical Name or Material Sodium pyruvate
EINECSNumber 204-024-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only