missing translation for 'onlineSavingsMsg'
Learn More

Sodium Pyruvate, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog No. p-5390950
Change view
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Wide Mouth Amber Glass Bottle
Catalog No. Quantity Packaging
02004229 100 g Wide Mouth Amber Glass Bottle
02004230 1 kg Wide Mouth Amber Glass Bottle
2 options

Catalog No. 02004229

Supplier: Avantor 335404

Only null left

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 113-24-6
Molecular Formula C3H3NaO3
Molecular Weight (g/mol) 110.044
InChI Key DAEPDZWVDSPTHF-UHFFFAOYSA-M
Synonym sodium pyruvate, pyruvic acid sodium salt, sodium 2-oxopropanoate, pyruvic acid, sodium salt, propanoic acid, 2-oxo-, sodium salt, sodium alpha-ketopropionate, pyruvate sodium, unii-pod38aif08, 2-oxopropanoic acid sodium salt, pod38aif08
PubChem CID 23662274
ChEBI CHEBI:50144
IUPAC Name sodium;2-oxopropanoate
SMILES CC(=O)C(=O)[O-].[Na+]

Specifications

Quantity 100 g
Formula Weight 110.05
Grade BAKER ANALYZED™ Reagent
Packaging Wide Mouth Amber Glass Bottle
Chemical Name or Material Sodium Pyruvate