missing translation for 'onlineSavingsMsg'
Learn More

Sodium Pyruvate, 99%, Spectrum™ Chemical

Catalog No. 18613840
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
25 kg
Packaging:
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613840 500 g Poly Pail
18613839 5 g Poly Pail
18613837 25 g Poly Pail
18613836 100 g Poly Pail
18613835 1 kg Poly Pail
18613838 25 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18613840 Supplier Spectrum Chemical Mfg Cor Supplier No. SO193500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 113-24-6

Specifications

CAS Max % 1
Physical Form Powder, Needles or Crystals
Quantity 500 g
Molecular Weight (g/mol) 110.05
Percent Purity 99%
Grade Reagent
Packaging Poly Pail
Chemical Name or Material Sodium Pyruvate

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.