missing translation for 'onlineSavingsMsg'
Learn More

Sodium Pyruvate, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor SO19325KGBL

 View more versions of this product

Catalog No. 18613838

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

113-24-6
25 kg
Reagent
1
99%
Fiber Drum

Chemical Identifiers

113-24-6
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.