missing translation for 'onlineSavingsMsg'
Learn More

Sodium Pyruvate, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor SO193100GM

 View more versions of this product

Catalog No. 18613836


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

113-24-6
100 g
Reagent
1
99%
Poly Bottle

Chemical Identifiers

113-24-6
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.