missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Sodium Pyruvate

Catalog No. P2256500G
Encompass
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2256500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P2256500G Supplier Sigma Aldrich Supplier No. P2256500G

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium Pyruvate

Specifications

CAS 101-84-8
Melting Point 25°C to 27°C
Density 1.073 g/mL at 25°C
Boiling Point 259°C
Molecular Formula C12H10O
Linear Formula (C6H5)2O
Quantity 500 g
MDL Number MFCD00003034
Synonym alpha-Ketopropionic acid sodium salt; 2-Oxopropanoic acid sodium salt; Pyruvic acid sodium salt
Molecular Weight (g/mol) 170.21 g/mol