missing translation for 'onlineSavingsMsg'
Learn More

Sodium pyrophosphate tetrabasic decahydrate, Puriss. p.a., ≥99%, Honeywell Fluka™

Catalog No. p-7071966
Change view
Click to view available options
Quantity:
2.5 kg
6 x 2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002264 2.5 kg
60045817 6 x 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6002264 Supplier Honeywell Chemicals Supplier No. 304112.5KG
Only null left

Puriss. p.a.,≥99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 13472-36-1
Molecular Formula H20Na4O17P2
Molecular Weight (g/mol) 446.05
InChI Key VZWGHDYJGOMEKT-UHFFFAOYSA-J
Synonym sodium pyrophosphate decahydrate, unii-iy3dkb96qw, tetrasodium pyrophosphate decahydrate, diphosphoric acid, tetrasodium salt, decahydrate, iy3dkb96qw, pyrophosphate sodium salt decahydrate, pyrophosphate inorganic, colostrum, sodium phosphate pyro, sodium pyrophosphate na4p2o7 decahydrate
PubChem CID 3084150
IUPAC Name tetrasodium;phosphonato phosphate;decahydrate
SMILES O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+]

Specifications

Quantity 2.5 kg
Linear Formula Na4P2O7 · 10H2O
MDL Number MFCD00149200
UN Number NONH for all modes of transport
Formula Weight 446.06g/mol
Percent Purity ≥99%
Grade puriss. p.a.
Chemical Name or Material Sodium Pyrophosphate Tetrabasic Decahydrate
Hazard Statement P280-P301 + P312 + P330-P305 + P351 + P338 + P310
Precautionary Statement H302-H318
EINECSNumber 231-767-1