missing translation for 'onlineSavingsMsg'
Learn More

Sodium Pyrophosphate, Decahydrate, Crystal, ACS, 99.0-103.0%, Spectrum™ Chemical

Catalog No. 18613830
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613830 125 g Poly Bottle
18613831 500 g Poly Bottle
18613832 2.5 kg Poly Bottle
18613833 12 kg Poly Pail
18613834 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18613830 Supplier Spectrum Chemical Mfg Cor Supplier No. S1420125GM
Only null left

Chemical Identifiers

CAS 13472-36-1
Molecular Formula H20Na4O17P2
Molecular Weight (g/mol) 446.05
InChI Key VZWGHDYJGOMEKT-UHFFFAOYSA-J
IUPAC Name tetrasodium decahydrate (phosphonatooxy)phosphonate
SMILES O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[Na+].[O-]P([O-])(=O)OP([O-])([O-])=O

Specifications

CAS Max % 1
pH 9.5 to 10.5
Quantity 125 g
Percent Purity 99 to 103%
Grade ACS
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.