missing translation for 'onlineSavingsMsg'
Learn More

Sodium Pyrophosphate Decahydrate, ACS Reagent, Reagents

Catalog No. p-200020868
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 lb.
10 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337689 10 lb.
01337688 10 kg
01337687 2.5 kg
01337686 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01337689 Supplier Labchem Inc Supplier No. C132458010F5
Only null left

Sodium Pyrophosphate Decahydrate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Boiling Point 93°C
Chemical Name or Material Sodium Pyrophosphate Decahydrate, ACS Reagent
Color Colorless/White
CAS 13472-36-1
Physical Form Solid
Quantity 10 lb.
Molecular Formula Na4P2O7, 10H2O
Synonym Tetrasodium Diphosphate Decahydrate
Molecular Weight (g/mol) 446.0552
Grade ACS
Packaging Poly Pail
Specific Gravity 1.82
Show More Show Less