missing translation for 'onlineSavingsMsg'
Learn More

Sodium pyrophosphate decahydrate, 97+%

CAS: 13472-36-1 | H20Na4O17P2 | 446.05 g/mol

Supplier:  Thermo Scientific Chemicals A161950B

 View more versions of this product

Catalog No. AAA161950B


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Sodium pyrophosphate decahydrate is used as a buffering agent, an emulsifier, a thickening agent, a food additive and a dispersing agent. It acts as a tartar control agent in tooth paste and dental floss and is used to remove calcium and magnesium from saliva, thereby avoiding its deposition on teeth. It is utilized in detergents to prevent deposition on cloth. It is useful in the preparation of an EDTA-sodium pyrophosphate extraction buffer for microcystin analysis of soil samples. In antimicrobial studies, it is used as a chelating agent.

Solubility
Soluble in water. Insoluble in alcohol and ammonia.

Notes
Incompatible with strong oxidizing agents
Specifications

Specifications

Sodium pyrophosphate decahydrate
13472-36-1
1000 g
Na4P2O7·10H2O
14,9240
Soluble in water. Insoluble in alcohol and ammonia.
O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+]
446.05
446.06 (265.90 Anhydrous)
Odorless
79.5°C
9.5 to 10.5
H20Na4O17P2
MFCD00149200
sodium pyrophosphate decahydrate, unii-iy3dkb96qw, tetrasodium pyrophosphate decahydrate, diphosphoric acid, tetrasodium salt, decahydrate, iy3dkb96qw, pyrophosphate sodium salt decahydrate, pyrophosphate inorganic, colostrum, sodium phosphate pyro, sodium pyrophosphate na4p2o7 decahydrate
VZWGHDYJGOMEKT-UHFFFAOYSA-J
tetrasodium;phosphonato phosphate;decahydrate
3084150
≥97%
1.82

Chemical Identifiers

13472-36-1
446.05
VZWGHDYJGOMEKT-UHFFFAOYSA-J
3084150
O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+]
H20Na4O17P2
MFCD00149200
sodium pyrophosphate decahydrate, unii-iy3dkb96qw, tetrasodium pyrophosphate decahydrate, diphosphoric acid, tetrasodium salt, decahydrate, iy3dkb96qw, pyrophosphate sodium salt decahydrate, pyrophosphate inorganic, colostrum, sodium phosphate pyro, sodium pyrophosphate na4p2o7 decahydrate
tetrasodium;phosphonato phosphate;decahydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 231-767-1

RTECSNumber : UX7350000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only