missing translation for 'onlineSavingsMsg'
Learn More

Sodium Pyrophosphate Anhydrous, Technical (Tetrasodium Pyrophosphate), Reagents

Catalog No. p-200021226
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336981 10 kg
01336980 2.5 kg
01336979 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01336981 Supplier Labchem Inc Supplier No. C232458010KG
Only null left

Sodium Pyrophosphate Anhydrous, Technical (Tetrasodium Pyrophosphate), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7722-88-5
Molecular Formula Na4O7P2
Molecular Weight (g/mol) 265.90
InChI Key FQENQNTWSFEDLI-UHFFFAOYSA-J
Synonym Tetrasodium Pyrophosphate
IUPAC Name tetrasodium (phosphonatooxy)phosphonate
SMILES [Na+].[Na+].[Na+].[Na+].[O-]P([O-])(=O)OP([O-])([O-])=O

Specifications

Melting Point 880°C
Color Colorless/White
Physical Form Solid
Quantity 10 kg
Grade Reagent
Packaging Poly Pail
Specific Gravity 2.53
Chemical Name or Material Sodium Pyrophosphate Anhydrous, Technical (Tetrasodium Pyrophosphate)