missing translation for 'onlineSavingsMsg'
Learn More

Sodium Pyrophosphate, Anhydrous, Powder, FCC, 95-100.5%, Spectrum™ Chemical

Catalog No. 18606764
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606764 500 g Poly Bottle
18606765 2.5 kg Poly Pail
18606766 12 kg Poly Pail
18606767 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18606764 Supplier Spectrum Chemical Mfg Cor Supplier No. S1419500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

S1419, 7722-88-5, Na4P2O7

Spectrum™ Chemical Sodium Pyrophosphate, Anhydrous, Powder, FCC it is used as a palatability enhancer. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7722-88-5
Molecular Formula Na4O7P2
Molecular Weight (g/mol) 265.90
MDL Number MFCD00003513
InChI Key FQENQNTWSFEDLI-UHFFFAOYSA-J
IUPAC Name tetrasodium (phosphonatooxy)phosphonate
SMILES [Na+].[Na+].[Na+].[Na+].[O-]P([O-])(=O)OP([O-])([O-])=O

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 95 to 100.5%
Grade FCC
Loss on Ignition 0.005
Packaging Poly Bottle