missing translation for 'onlineSavingsMsg'
Learn More

Sodium Pyridine-2-carboxylate 98.0+%, TCI America™

Catalog No. P042425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P042425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P042425G Supplier TCI America Supplier No. P042425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Laboratory reagent used as a building block for chemical synthesis.

Chemical Identifiers

CAS 57665-05-1
Molecular Formula C6H4NNaO2
Molecular Weight (g/mol) 145.093
MDL Number MFCD00070517
InChI Key RLEBKHAOAHYZHT-UHFFFAOYSA-M
Synonym Pyridine-2-carboxylic Acid Sodium Salt, Picolinic Acid Sodium Salt, Sodium Picolinate
PubChem CID 23668608
IUPAC Name sodium;pyridine-2-carboxylate
SMILES C1=CC=NC(=C1)C(=O)[O-].[Na+]

Specifications

Color White
Quantity 25 g
Formula Weight 145.09
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Pyridine-2-carboxylate