Learn More
Sodium Potassium Tartrate Tetrahydrate (Crystalline/ ACS Grade), Fisher Chemical™
Supplier: Thermo Fisher Scientific S38710
Specifications
| Sodium Potassium Tartrate Tetrahydrate | |
| >100°C | |
| White | |
| 437.22°C | |
| Poly Pail | |
| MFCD00150989 | |
| potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j | |
| VZOPRCCTKLAGPN-UHFFFAOYSA-L | |
| potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate | |
| 2724148 | |
| 99 to 102% | |
| Powder or Solid |
| 6381-59-5 | |
| 70°C | |
| 5.5 to 8.5 | |
| 99 to 102% | |
| C4H12KNaO10 | |
| 10 kg | |
| 0.005% max. Insoluble Matter | |
| C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+] | |
| 282.218 | |
| 282.22 | |
| ACS |
Chemical Identifiers
| 6381-59-5 | |
| 282.218 | |
| VZOPRCCTKLAGPN-UHFFFAOYSA-L | |
| 2724148 | |
| C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+] |
| C4H12KNaO10 | |
| MFCD00150989 | |
| potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j | |
| potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate |
Safety and Handling
CAUTION!
Emergency Overview
May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .
NFPA
Health:1
Flammability:1
Instability:0