missing translation for 'onlineSavingsMsg'
Learn More

Sodium Potassium Tartrate, Tetrahydrate, ACS Grade, 99.0 to 102.0%, LabChem™

Catalog No. LC247851
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC247851 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC247851 Supplier LabChem Supplier No. LC247851

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6381-59-5
Molecular Formula C4H12KNaO10
Molecular Weight (g/mol) 282.218
InChI Key VZOPRCCTKLAGPN-UHFFFAOYSA-L
Synonym potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j
PubChem CID 2724148
IUPAC Name potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate
SMILES C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+]

Specifications

Melting Point 158°C
Color White
CAS Max % 100
Physical Form Crystals
Quantity 500 g
Linear Formula NaKC4H4O6·4H2O
Solubility Information Soluble in water
Formula Weight 282.23
Percent Purity 99.0 to 102.0%
Grade ACS
Identification Passes Test
Packaging Poly Bottle
Decomposition Information Sodium oxide; Carbon monoxide; Carbon dioxide
Chemical Name or Material Sodium Potassium Tartrate, Tetrahydrate
Show More Show Less
Hazard Statement GHS H Statement
Product is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
EINECSNumber 206-156-8
Recommended Storage Room Temperature