missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphite dibasic pentahydrate, 98%

Catalog No. p-4813662
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC428502500 250 g Glass bottle
AC428500010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC428502500 Supplier Thermo Scientific Chemicals Supplier No. 428502500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13517-23-2 ∣ HNa2O3P·5H2O ∣ 216.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13517-23-2
Molecular Formula HNa2O3P·5H2O
Molecular Weight (g/mol) 216.02
Synonym sodium phosphite dibasic pentahydrate, di-sodium hydrogen phosphite, di-sodium hydrogen phosphite pentahydrate, disodium pentahydrate hydrogen phosphite, 2na.ho3p.5h2o, di-sodiumhydrogenphosphitepentahydrate, disodium hydrogenphosphite pentahydrate, etidronate disodium related compound a, united states pharmacopeia usp reference standard

Specifications

Physical Form Crystals or Needles
Quantity 250 g
Assay Percent Range 98%
Merck Index 15,8793
Solubility Information Solubility in water: soluble
Formula Weight 216.02
Percent Purity 98%
Grade Ungraded
Packaging Glass bottle
Chemical Name or Material Sodium phosphite dibasic pentahydrate

RUO – Research Use Only