missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate, tribasic, pure, anhydrous

Catalog No. p-3777076
Change view
Click to view available options
Quantity:
1 kg
10 kg
Packaging:
Fiber Drum
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC389810010 1 kg Plastic Bottle
AC389810100 10 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC389810010 Supplier Thermo Scientific Chemicals Supplier No. 389810010
Only null left

CAS: 7601-54-9 | Na3O4P | 163.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7601-54-9
Molecular Formula Na3O4P
Molecular Weight (g/mol) 163.94
MDL Number MFCD00003510
InChI Key RYFMWSXOAZQYPI-UHFFFAOYSA-K
Synonym trisodium phosphate, sodium orthophosphate, tromete, tribasic sodium phosphate, phosphoric acid, trisodium salt, oakite, sodium phosphate, anhydrous, trisodium orthophosphate, phosphoric acid trisodium salt, trinatriumphosphat
PubChem CID 24243
ChEBI CHEBI:37583
IUPAC Name trisodium;phosphate
SMILES [O-]P(=O)([O-])[O-].[Na+].[Na+].[Na+]

Specifications

Color White
Physical Form Crystalline Powder
Quantity 1 kg
Linear Formula Na3PO4
Merck Index 15, 8792
Solubility Information Solubility in water: 88g/L (20°C)
Formula Weight 163.94
Percent Purity ≥96.0%
Grade Pure
Packaging Plastic Bottle
Chemical Name or Material Sodium phosphate, tribasic, Anhydrous, Pure
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:
EINECSNumber 231-509-8

RUO – Research Use Only