missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate Tribasic Dodecahydrate, ACS Reagent (Trisodium Phosphate Dodecahydrate), Reagents

Catalog No. 01337692
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337692 10 kg
01337691 2.5 kg
01337690 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01337692 Supplier Labchem Inc Supplier No. C132450010KG
Only null left

Sodium Phosphate Tribasic Dodecahydrate, ACS Reagent (Trisodium Phosphate Dodecahydrate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Sodium Phosphate Tribasic Dodecahydrate, ACS Reagent (Trisodium Phosphate Dodecahydrate)
Color Colorless/White
CAS 10101-89-0
Physical Form Solid
Quantity 10 kg
Molecular Formula Na3PO4, 12H2O
Synonym Trisodium Phosphate Dodecahydrate
Molecular Weight (g/mol) 380.124
Grade ACS
Packaging Poly Pail
Specific Gravity 1.6
Show More Show Less